EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Pharmacokinetic properties of the synthetic cannabinoid JWH-018 and of its metabolites in serum after inhalation.
PMID 28365515
Abstract
Each year, synthetic cannabinoids are occurring in high numbers in the illicit drug market, but data on their pharmacology and toxicology are scarcely available. Therefore, a pilot study was performed to assess adverse effects of JWH-018, which is one of the oldest and best known synthetic cannabinoids. Six subjects inhaled smoke from 2 and 3mg JWH-018. The drug and nine of its metabolites were analyzed in their blood samples taken during the following 12h by liquid chromatography-mass spectrometry (LC-MSMS). The maximum concentration of JWH-018 reached 2.9-9.9ng/ml after inhalation and markedly decreased during the next 1.5h, followed by a multiexponential decline (t
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
219878
4-Hydroxyindole, 99%
C8H7NO