EMAIL THIS PAGE TO A FRIEND
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Surface Properties and MC3T3-E1 Cell Response of Cortical Bone Allografts Modified with Low-Concentration Phosphoric Acid.
PMID 28365682
Abstract
This experimental study aimed to evaluate the effect of low-concentration phosphoric acid on the surface structure of cortical allografts. Allogenic cortical bones were obtained from femurs and tibias of New Zealand white rabbits. The bones were modified by treatment with various concentrations of phosphoric acid (10%, 20% or 30%) for 10, 30 or 60 minutes, then evaluated by the following methods: 3-(4,5-Dimethyl-2-thiazolyl)-2,5-diphenyl-2H-tetrazolium bromide (MTT) and LIVE/DEAD assay, alkaline phosphatase (ALP) assay, biomechanical properties testing, contact angle detection, quantitative real-time polymerase chain reaction (Q-PCR), western blotting and scanning electron microscopy (SEM). Compared with the other groups, the group modified with 10% H3PO4 for 10 minutes had lower cytotoxicity according to MTT and LIVE/DEAD assays, higher hydrophilicity in the contact angle detection test and greater stability in the biomechanical properties test. Moreover, an up-regulation of osteopontin (OPN) in bones modified with 10% H3PO4 was observed by Q-PCR and western blotting. In addition, ALP assay and SEM showed that surface porosity and osteoinductivity were increased in the group modified with 10% H3PO4. Low-concentration phosphoric acid may be a potential method for surface modification of cortical allografts. Further animal experiments and animal infection model studies are required to validate the efficacy of surface-modified cortical allografts to repair large segmental bone defects.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0332
Edelfosine, ≥95% (HPLC)
C27H58NO6P