EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
JTP-117968, a novel selective glucocorticoid receptor modulator, exhibits improved transrepression/transactivation dissociation.
PMID 28366807
Abstract
Classic glucocorticoids that have outstanding anti-inflammatory effects are still widely prescribed for the treatment of various inflammatory and autoimmune diseases. Conversely, glucocorticoids cause numerous unwanted side effects, particularly systemically dosed glucocorticoids. Therefore, selective glucocorticoid receptor modulator (SGRM), which maintains beneficial anti-inflammatory effects while reducing the occurrence of side effects, is one of the most anticipated drugs. However, there have been no SGRMs marketed to date. The assumption is that there are two major mechanisms of action of glucocorticoids via glucocorticoid receptors, transrepression (TR) and transactivation (TA). In general, the anti-inflammatory effects of glucocorticoids are mostly mediated through TR, while the side effects associated with glucocorticoids are largely caused by TA. We started to evaluate novel orally available SGRMs that maintain anti-inflammatory effects while minimizing adverse effects by favoring TR over TA. Based on this evaluation, we discovered JTP-117968, (4b'S,7'R,8a'S)-4b'-benzyl-7'-hydroxy-N-(2-methylpyridin-3-yl)-7'-(trifluoromethyl)-4b',6',7',8',8a',10'-hexahydro-5'H-spiro[cyclopropane-1,9'-phenanthrene]-2'-carboxamide, a non-steroidal SGRM. JTP-117968 has partial TR activity, but exhibits extremely low TA activity. The maximum TR efficacy of JTP-117968 was comparable to its structural analogue, PF-802, (4bS,7R,8aR)-4b-Benzyl-7-hydroxy-N-(2-methylpyridin-3-yl)-7-(trifluoromethyl)-4b,5,6,7,8,8a,9,10-octahydrophenanthrene-2-carboxamide, which is the active form of Fosdagrocorat that has been developed clinically as a first-in-class orally available SGRM. Remarkably, the TA activity of JTP-117968 was much weaker than PF-802 not only in in vitro assays, but also in in vivo mice experiments. These findings indicate that JTP-117968 exhibits improved TR/TA dissociation because the compound has significantly lower TA activity compared with an already reported SGRM. Therefore, JTP-117968 is expected to be a useful compound for evaluating ideal SGRMs in the future.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56924
Tricosanoic acid tryptamide, analytical standard
C33H56N2O