EMAIL THIS PAGE TO A FRIEND
Journal of Asian natural products research
A new pre-column derivatization for valienamine and beta-valienamine using o-phthalaldehyde to determine the epimeric purity by HPLC and application of this method to monitor enzymatic catalyzed synthesis of beta-valienamine.
PMID 28367638
Abstract
Valienamine and β-valienamine are representative C
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
735914
Octylphosphonic acid, 97%
C8H19O3P