EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Effects of Salt Concentrations and Nitrogen and Phosphorus Starvations on Neutral Lipid Contents in the Green Microalga Dunaliella tertiolecta.
PMID 28368591
Abstract
Dunaliella tertiolecta, a halotolerant alga, can accumulate large amounts of neutral lipid, which makes it a potential biodiesel feedstock. In this study, neutral lipids of D. tertiolecta induced by different salinities or N or P starvation were analyzed by thin-layer chromatography (TLC), flow cytometry (FCM), and confocal laser scanning microscopy (CLSM). High salinities or N or P starvation resulted in a decrease in cell growth and chlorophyll contents of D. tertiolecta. Neutral lipid contents increased markedly after 3-7 days of N starvation or at low NaCl concentrations (0.5-2.0 M). N starvation had a more dramatic effect on the neutral lipid contents of D. tertiolecta than P starvation. Four putative ME isozymes in different conditions can be detected by using isozyme electrophoresis. Two alternative acetyl-CoA producers, ACL and ACS genes, were up-regulated under low salinities and N starvation. It was suggested that low salinities and N starvation are considered efficient ways to stimulate lipid accumulation in D. tertiolecta.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00538
Chlorophyll b, analytical standard
C55H70MgN4O6