EMAIL THIS PAGE TO A FRIEND
Roczniki Panstwowego Zakladu Higieny
[Quantitative changes in tyrosine levels in experimental Ferbam 76 poisoning. IV. Levels of free and bound tyrosine in the organs of experimental animals].
PMID 2836943
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45496
Ferbam, PESTANAL®, analytical standard
C9H18FeN3S6