EMAIL THIS PAGE TO A FRIEND
Chemphyschem : a European journal of chemical physics and physical chemistry
One-Pot Hybrid SnO
PMID 28371061
Abstract
The localized in situ formation of tin dioxide (SnO
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
345164
Tin(II) acetate
C4H6O4Sn