EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Atrial natriuretic factor inhibits phosphate uptake in opossum kidney cells: as a model of renal proximal tubules.
PMID 2837187
Abstract
The cultured renal cell, an opossum kidney (OK) cell line, which contains several features characteristic of proximal tubular cells, was utilized to examine the direct effects of atrial natriuretic factor (ANF) and cyclic GMP (cGMP) on phosphate uptake. ANF at 2 x 10(-7) M significantly inhibited phosphate uptake by 10.1% of control (P less than 0.01). Incubation of the cells with ANF (10(-8) to 10(-6) M) resulted in an increment of intracellular cGMP in a dose dependent fashion. Exogenous addition of 8-bromo-cGMP (10(-4) M) also significantly inhibited phosphate uptake by 14.6%. These results suggest that ANF directly inhibits phosphate transport in renal proximal tubular cells, probably through stimulation of cGMP production.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
OK, 91021202