EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Parathyroid hormone controls paracellular Ca
PMID 28373577
Abstract
Renal Ca
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4151
Sodium decanoate, ≥98%
C10H19NaO2