EMAIL THIS PAGE TO A FRIEND
Scientific reports
A Gate Hinge Controls the Epithelial Calcium Channel TRPV5.
PMID 28374795
Abstract
TRPV5 is unique within the large TRP channel family for displaying a high Ca