EMAIL THIS PAGE TO A FRIEND
European journal of haematology
Genomewide association study on monoclonal gammopathy of unknown significance (MGUS).
PMID 28375557
Abstract
To identify germ line variants contributing to the development of monoclonal gammopathy of undetermined significance (MGUS), an asymptomatic premalignant precursor for multiple myeloma (MM). We conducted the first genomewide association study (GWAS) on MGUS on 243 German cases with a replication on 294 Czech cases. Identified loci were further analyzed in 1508 German MM patients. New MM loci recently reported in a meta-analysis were also tested in the MGUS GWAS. In GWAS, we identified 10 loci contributing to development of MGUS at P-value threshold of 10 These data allow a cautious first proposal for a germ line architecture of MGUS with links to leukemia and autoimmune conditions, the latter agreeing with a family study showing clustering of MGUS with autoimmune diseases.