EMAIL THIS PAGE TO A FRIEND
The EMBO journal
An aberrant phase transition of stress granules triggered by misfolded protein and prevented by chaperone function.
PMID 28377462
Abstract
Stress granules (SG) are membrane-less compartments involved in regulating mRNAs during stress. Aberrant forms of SGs have been implicated in age-related diseases, such as amyotrophic lateral sclerosis (ALS), but the molecular events triggering their formation are still unknown. Here, we find that misfolded proteins, such as ALS-linked variants of SOD1, specifically accumulate and aggregate within SGs in human cells. This decreases the dynamics of SGs, changes SG composition, and triggers an aberrant liquid-to-solid transition of