EMAIL THIS PAGE TO A FRIEND
Frontiers in physiology
Tissue Nonspecific Alkaline Phosphatase (TNAP) Regulates Cranial Base Growth and Synchondrosis Maturation.
PMID 28377728
Abstract
Hypophosphatasia is a rare heritable disorder caused by inactivating mutations in the gene (