EMAIL THIS PAGE TO A FRIEND
Environmental science and pollution research international
Foam-forming bacteria in activated sludge effectively reduced by rotifers in laboratory- and real-scale wastewater treatment plant experiments.
PMID 28378311
Abstract
Lecane inermis rotifers were shown to diminish sludge bulking due to their ability to ingest the filamentous bacteria in activated sludge. To determine if rotifers are also able to control branched actinomycetes, we investigated three other Lecane species (Monogononta). In a week-long experiment, only Lecane tenuiseta significantly reduced the density of Microthrix parvicella and Type 0092 filaments, but in a 2-week experiment, actinomycetes were significantly reduced by most of the tested monogonont rotifers: L. inermis, Lecane decipiens and Lecane pyriformis. Rotifers L. inermis originating from the mass culture were artificially introduced into real-scale wastewater treatment plant (WWTP) in two series. The WWTP was monitored for 1 year. Rotifer inoculation resulted in diminishing of M. parvicella and actinomycete abundance. The experiments showed that different species of rotifers vary in their effectiveness at limiting various types of filamentous organisms. This is the first report demonstrating that one of the most troublesome bacteria, branched actinomycetes, which cause heavy foaming in bioreactors, can be controlled by rotifers. Knowledge of the consumers of filamentous bacteria that inhabit activated sludge could help WWTP operators overcome bulking and foaming through environmentally friendly methods.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
88071
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
C44H30N4