EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Cell physiology
Balance between
PMID 28381519
Abstract
Nitric oxide (NO) contributes to myogenesis by regulating the transition between myoblast proliferation and fusion through cGMP signaling. NO can form