EMAIL THIS PAGE TO A FRIEND
Frontiers in microbiology
Notable Increasing Trend in Azole Non-susceptible
PMID 28382028
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32103
Posaconazole, VETRANAL, analytical standard
C37H42F2N8O4