EMAIL THIS PAGE TO A FRIEND
Molecular oral microbiology
The two-component system VicRK regulates functions associated with Streptococcus mutans resistance to complement immunity.
PMID 28382721
Abstract
Streptococcus mutans, a dental caries pathogen, can promote systemic infections upon reaching the bloodstream. The two-component system (TCS) VicRK