EMAIL THIS PAGE TO A FRIEND
Scientific reports
p38α MAPK regulates proliferation and differentiation of osteoclast progenitors and bone remodeling in an aging-dependent manner.
PMID 28382965
Abstract
Bone mass is determined by the balance between bone formation, carried out by mesenchymal stem cell-derived osteoblasts, and bone resorption, carried out by monocyte-derived osteoclasts. Here we investigated the potential roles of p38 MAPKs, which are activated by growth factors and cytokines including RANKL and BMPs, in osteoclastogenesis and bone resorption by ablating p38α MAPK in LysM+monocytes. p38α deficiency promoted monocyte proliferation but regulated monocyte osteoclastic differentiation in a cell-density dependent manner, with proliferating p38α
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU092951 MISSION® esiRNA, esiRNA human CREB1 (esiRNA1)