EMAIL THIS PAGE TO A FRIEND
European journal of immunology
Monocytic myeloid-derived suppressor cells regulate T-cell responses against vaccinia virus.
PMID 28383204
Abstract
Vaccinia virus (VV) can potently activate NK- and T-cell responses, leading to efficient viral control and generation of long-lasting protective immunity. However, immune responses against viral infections are often tightly controlled to avoid collateral damage and systemic inflammation. We have previously shown that granulocytic myeloid-derived suppressor cells (g-MDSCs) can suppress the NK-cell response to VV infection. It remains unknown what regulates T-cell responses to VV infection in vivo. In this study, we first showed that monocytic MDSCs (m-MDSCs), but not g-MDSCs, from VV-infected mice could directly suppress CD4
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M7033
NG-Methyl-L-arginine acetate salt, ≥98% (TLC)
C7H16N4O2 · C2H4O2