EMAIL THIS PAGE TO A FRIEND
Scientific reports
Single Molecule Translation Imaging Visualizes the Dynamics of Local β-Actin Synthesis in Retinal Axons.
PMID 28386060
Abstract
Local mRNA translation occurs in growing axons enabling precise control of the proteome in response to signals. To measure quantitatively the spatiotemporal dynamics of protein synthesis in growth cones, we further developed a technique for single molecule translation imaging (SMTI). We report that Netrin-1 triggers a burst of β-actin synthesis at multiple non-repetitive sites, particularly in the periphery. The response is remarkably rapid starting within 20 seconds of cue application.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0040
PP1, ≥98% (HPLC)
C16H19N5