EMAIL THIS PAGE TO A FRIEND
Environmental science & technology
Isomerization of Second-Generation Isoprene Peroxy Radicals: Epoxide Formation and Implications for Secondary Organic Aerosol Yields.
PMID 28388039
Abstract
We report chamber measurements of secondary organic aerosol (SOA) formation from isoprene photochemical oxidation, in which radical concentrations were systematically varied and the molecular composition of semi- to low-volatility gases and SOA were measured online. Using a detailed chemical kinetics box model, we find that to explain the behavior of low-volatility products and SOA mass yields relative to input H
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11202
Adenosine 2′-phosphate 5′-phosphosulfate lithium salt, ≥85% (HPLC)
C10H15N5O13P2S · xLi+