EMAIL THIS PAGE TO A FRIEND
Journal of ethnopharmacology
Compatibility of Tanshinone IIA and Astragaloside IV in attenuating hypoxia-induced cardiomyocytes injury.
PMID 28389356
Abstract
Herbal medicines including Tanshinone IIA (TanIIA) and Astragaloside IV (AsIV) are widely used in Asia as therapeutic agents for cardiovascular diseases, due to their complementary roles and shared properties based on the theory of traditional Chinese medicine and pharmacological researches. However, the underlying pathological mechanisms for their efficacy are still unclear. In addition, the compatibility or incompatibility of the herbal medicines when administered with other herbal remedies or with prescription drugs is unknown. We aimed to investigate the compatibility of TanIIA and AsIV in protecting cardiomyocytes against hypoxia-induced injury. Cultured cardiomyocytes were stimulated in hypoxia condition, in the absence or presence of the two herbal compounds, TanIIA and AsIV. Indicators were determined by cytotoxicity assay, quantitative PCR, ELISA, flow cytometry assay, immunofluorescence staining and western blot. Either TanIIA alone or the combined herbal compounds inhibited hypoxia-triggered chemokines production including CCL2/5/19, CXCL2 and Transwell assay-indicated monocyte/macrophage recruitment, cytokines production including TNF-α and IL-6. While AsIV alone or the combined herbal compounds attenuated hypoxia-induced cell apoptosis indicated by decreased Annexin V Taken together, these data suggest a compatibility of TanIIA and AsIV in protecting cardiomyocyte against hypoxia-induced injury.