Reproduction, fertility, and development
Supplementation of culture medium with L-carnitine improves the development and cryotolerance of in vitro-produced porcine embryos.
PMID 28390470
Porcine oocytes and embryos contain substantial amounts of lipid, with little known regarding its metabolic role during development. This study investigated the role of lipid metabolism and the interaction between carbohydrate and lipid substrates in porcine embryos. Following in vitro fertilisation, presumptive zygotes were transferred to culture medium supplemented with L-carnitine, a co-factor required for the metabolism of fatty acids. In porcine zygote medium-3 (PZM-3), which contains pyruvate and lactate, 3mM L-carnitine was the only dose that improved cleavage rates compared with the control. In the absence of carbohydrates, all doses of L-carnitine from 1.5 to 12mM increased cleavage rates compared with the control. Culture in a PZM-3-based sequential media system (Days 0-3: pyruvate and lactate; Days 4-7: glucose) significantly increased blastocyst cell numbers compared with culture in standard PZM-3. Supplementing PZM-3 with 3mM L-carnitine produced blastocysts with cell numbers equivalent to those obtained in the sequential media system. After vitrification, the post-warming survival rates of blastocysts obtained in media supplemented with 3mM L-carnitine were significantly greater than those of blastocysts obtained in standard PZM-3. In conclusion, L-carnitine supplementation improved embryo development when the medium contained pyruvate and lactate or was lacking carbohydrates completely, indicating a role for fatty-acid metabolism when the embryo's requirements for carbohydrates are not adequately met.
Related Materials
Product #
Molecular Formula
Add to Cart
Hypotaurine, ≥98% (TLC)