EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Relationship of angiotensin I-converting enzyme (ACE) and bradykinin B2 receptor (BDKRB2) polymorphism with diabetic nephropathy.
PMID 28390948
Abstract
To determine whether ACE We provide first evidence indicating the causation between ACE DD or B2R+9bp genotype and the increased risk for diabetic nephropathy, broadening our horizon about the role of genetic modulators in this disease.