EMAIL THIS PAGE TO A FRIEND
Forensic science international. Genetics
Development and validation of a novel multiplexed DNA analysis system, InnoTyper
PMID 28391141
Abstract
We report here a novel multiplexed DNA analysis system consisting of 20 Alu markers and Amelogenin for analysis of highly degraded forensic biological samples. The key to the success of the system in obtaining results from degraded samples is the primer design yielding small amplicon size (60-125bp) for all 20 markers. The markers included in the InnoTyper
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H3281
Hematin porcine
C34H33FeN4O5