EMAIL THIS PAGE TO A FRIEND
Environmental science & technology
Predicting Reactive Intermediate Quantum Yields from Dissolved Organic Matter Photolysis Using Optical Properties and Antioxidant Capacity.
PMID 28391701
Abstract
The antioxidant capacity and formation of photochemically produced reactive intermediates (RI) was studied for water samples collected from the Florida Everglades with different spatial (marsh versus estuarine) and temporal (wet versus dry season) characteristics. Measured RI included triplet excited states of dissolved organic matter (
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W249106
Furfuryl alcohol, ≥97%, FG
C5H6O2