European journal of medicinal chemistry
Novel 2-pyridinecarboxaldehyde thiosemicarbazones Ga(III) complexes with a high antiproliferative activity by promoting apoptosis and inhibiting cell cycle.
PMID 28395152
Two types of 2-pyridinecarboxaldehyde thiosemicarbazones Ga(III) complexes, which are 2:1 and 1:1 ligand/Ga(III) complexes, were synthesized and determined by X-ray single crystal diffraction. The antiproliferative activity of these Ga(III) complexes have been examined to illuminate the structure-activity relationships essential to form Ga(III) complexes with remarkable anticancer activity. In addition, Ga(III) complexes where the metal/ligand ratio was 1:1 (C4) had observably higher antiproliferative activity than 1:2 (C3). Ga(III) complexes caused a marked increase of caspase-3 and 9 activity in NCI-H460 cells compared to the metal free ligand. Caspase activation was somewhat mediated by the release of Cyt C from mitochondria after incubation with selected agents. Both types of Ga(III) complexes showed more effective in inhibition of the G1/S transition than the ligand alone.