EMAIL THIS PAGE TO A FRIEND
Journal of molecular and cellular cardiology
Acetylcholine attenuated TNF-α-induced intracellular Ca
PMID 28395930
Abstract
The endoplasmic reticulum (ER) forms discrete junctions with the plasma membrane (PM) that play a critical role in the regulation of Ca
Related Materials