Cancer research
Extracellular Matrix/Integrin Signaling Promotes Resistance to Combined Inhibition of HER2 and PI3K in HER2(+) Breast Cancer.
PMID 28396358
PIK3CA mutations are associated with resistance to HER2-targeted therapies. We previously showed that HER2(+)/PIK3CA(H1047R) transgenic mammary tumors are resistant to the HER2 antibodies trastuzumab and pertuzumab but respond to PI3K inhibitor buparlisib (TPB). In this study, we identified mechanisms of resistance to combined inhibition of HER2 and PI3K. TPB-resistant tumors were generated by treating HER2(+)/PIK3CA(H1047R) tumor-bearing mice long term with the drug combination. RNA sequencing of TPB-resistant tumors revealed that extracellular matrix and cell adhesion genes, including collagen II (Col2a1), were markedly upregulated, accompanied by activation of integrin β1/Src. Cells derived from drug-resistant tumors were sensitive to TBP when grown in vitro, but exhibited resistance when plated on collagen or when reintroduced into mice. Drug resistance was partially reversed by the collagen synthesis inhibitor ethyl-3,4-dihydroxybenzoate. Inhibition of integrin β1/Src blocked collagen-induced resistance to TPB and inhibited growth of drug-resistant tumors. High collagen II expression was associated with significantly lower clinical response to neoadjuvant anti-HER2 therapy in HER2(+) breast cancer patients. Overall, these data suggest that upregulation of collagen/integrin/Src signaling contributes to resistance to combinatorial HER2 and PI3K inhibition. Cancer Res; 77(12); 3280-92. ©2017 AACR.
Related Materials
Product #
Molecular Formula
Add to Cart
Protocatechuic acid ethyl ester, 97%