EMAIL THIS PAGE TO A FRIEND
Drug metabolism and disposition: the biological fate of chemicals
Human Enterocytes as an In Vitro Model for the Evaluation of Intestinal Drug Metabolism: Characterization of Drug-Metabolizing Enzyme Activities of Cryopreserved Human Enterocytes from Twenty-Four Donors.
PMID 28396528
Abstract
We report in this work successful isolation and cryopreservation of enterocytes from human small intestine. The enterocytes were isolated by enzyme digestion of the intestinal lumen, followed by partial purification via differential centrifugation. The enterocytes were cryopreserved directly after isolation without culturing to maximize retention of in vivo drug-metabolizing enzyme activities. Post-thaw viability of the cryopreserved enterocytes was consistently over 80% based on trypan blue exclusion. Cryopreserved enterocytes pooled from eight donors (four male and four female) were evaluated for their metabolism of 14 pathway-selective substrates: CYP1A2 (phenacetin hydroxylation), CYP2A6 (coumarin 7-hydroxylation), CYP2B6 (bupropion hydroxylation), CYP2C8 (paclitaxel 6
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
UC205
Dextrorphan
C17H23NO