EMAIL THIS PAGE TO A FRIEND
Antioxidants & redox signaling
Beyond a Gasotransmitter: Hydrogen Sulfide and Polysulfide in Cardiovascular Health and Immune Response.
PMID 28398086
Abstract
Hydrogen sulfide (H In light of the recent understanding of oxidized sulfide metabolite formation and reactivity, comparatively few studies have been reported comparing cellular biological and in vivo effects of H Additional studies regarding persulfide and polysulfide formation and molecular reactions are needed in nearly all aspects of biology to better understand how sulfide metabolites contribute to key chemical biology reactions involved in cardiovascular health and immune responses. Antioxid. Redox Signal. 27, 634-653.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3633
Cystathionine, ≥90% (HPLC)
C7H14N2O4S