EMAIL THIS PAGE TO A FRIEND
Molecular carcinogenesis
Ski regulates Smads and TAZ signaling to suppress lung cancer progression.
PMID 28398634
Abstract
Ski, the transforming protein of the avian Sloan-Kettering retrovirus, displays both pro- and anti-oncogenic activities in human cancer. The mechanisms underlying these conflicting observations have not been fully understood. Herein, we investigated the mechanism underlying the tumor suppressor activity of Ski. To investigate the effect of Ski re-activation on TGF-β and Hippo/TAZ pathway, we measured its effect on the endogenous Smad target genes (PAI-1 and P15