EMAIL THIS PAGE TO A FRIEND
Life sciences
Phosphatidylinositol 3-kinase inhibition induces vasodilator effect of sevoflurane via reduction of Rho kinase activity.
PMID 28400117
Abstract
This study was aimed to examine whether a volatile anesthetic sevoflurane in clinical doses reduces vasoconstriction under the inhibition of phosphatidylinositol 3-kinase (PI3K) in the rat and human arteries and whether the intravenous administration of the PI3K inhibitor decreases blood pressure in rats under the sevoflurane inhalation. Rat arteries (n=5-6) and human omental arteries (n=5-6) were subjected to isometric force recordings and western immunoblotting for Rho kinase, mitogen-activated protein kinase, and protein kinase C. Some arteries were incubated with sevoflurane (1.5% or 3%), a selective PI3K inhibitor LY294002 (3×10 Sevoflurane with LY294002, but not sevoflurane or LY294002 solely, inhibited the phenylephrine-induced contraction (32% to 52% decrease at phenylephrine [3×10 Clinical sevoflurane doses with PI3K inhibition reduce the contraction of rat and human arteries ex vivo resulting from Rho kinase inhibition, and systemic blood pressure of rats in vivo. These results suggest that sevoflurane potentially causes vasodilation and hypotension in patients receiving anti-cancer therapy that inhibits PI3K.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G5251
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
C3H7O6P