EMAIL THIS PAGE TO A FRIEND
The Plant cell
ELF18-INDUCED LONG-NONCODING RNA Associates with Mediator to Enhance Expression of Innate Immune Response Genes in Arabidopsis.
PMID 28400491
Abstract
The plant immune response is a complex process involving transcriptional and posttranscriptional regulation of gene expression. Responses to plant immunity are initiated upon the perception of pathogen-associated molecular patterns, including peptide fragment of bacterial flagellin (flg22) or translation elongation factor Tu (elf18). Here, we identify an
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D134406
3′,5′-Dimethoxy-4′-hydroxyacetophenone, 97%
C10H12O4