EMAIL THIS PAGE TO A FRIEND
Cell biology international
Formation of the IGF1R/CAV1/SRC tri-complex antagonizes TRAIL-induced apoptosis in gastric cancer cells.
PMID 28403518
Abstract
Lipid rafts provide a biological platform for apoptosis induced by tumor necrosis factor-related apoptosis-inducing ligand (TRAIL). We previously reported that insulin-like growth factor 1 receptor (IGF1R) translocation into lipid rafts helped to explain TRAIL resistance. However, it was not clear whether TRAIL resistance was caused by the interaction of IGF1R with caveolin-1 (CAV1) and the non-receptor tyrosine kinase SRC in lipid rafts of gastric cancer cells. Here, we observed high IGF1R expression in TRAIL-resistant gastric cancer cells, and showed that IGF1R combined with both CAV1 and SRC in a native complex. TRAIL was shown to promote the formation of the IGF1R/CAV1/SRC tri-complex and the activation of these three molecules. Knockdown of IGF1R or CAV1 or inhibition of SRC activity reduced the formation of this tri-complex and enhanced TRAIL-induced apoptosis. Furthermore, the overexpression of microRNA-194 reversed TRAIL resistance by reducing IGF1R expression. In summary, TRAIL increased formation of the IGF1R/CAV1/SRC tri-complex and the activation of downstream survival pathways, leading to TRAIL resistance in gastric cancer cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0752
Adenosine 5′-diphosphoribose sodium salt, ≥93%
C15H23N5O14P2