EMAIL THIS PAGE TO A FRIEND
Natural product research
Two new cytochalasan derivatives from Chaetomium globosum SNSHI-5, a fungus derived from extreme environment.
PMID 28403641
Abstract
Two new cytochalasan derivatives, isochaetoglobosin D
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
774243
1,3-Propylene sulfite, 99%
C3H6O3S