EMAIL THIS PAGE TO A FRIEND
Molecular biology of the cell
Wound-induced Ca
PMID 28404746
Abstract
Damage-associated molecular patterns (DAMPs) are critical mediators of information concerning tissue damage from damaged cells to neighboring healthy cells. ATP acts as an effective DAMP when released into extracellular space from damaged cells. Extracellular ATP receptors monitor tissue damage and activate a Ca
Related Materials