EMAIL THIS PAGE TO A FRIEND
Oncotarget
Gambogic acid suppresses cancer invasion and migration by inhibiting TGFβ1-induced epithelial-to-mesenchymal transition.
PMID 28404892
Abstract
The epithelial-to-mesenchymal transition (EMT) contributes to the disruption of cell-cell junctions and imbues cancer cells with invasive and migratory properties. In this study, we investigated the effect of gambogic acid, a xanthone extracted from the resin of Garciania hanburyi, on transforming growth factor β1 (TGFβ1)-induced EMT. Gambogic acid inhibited the invasion and migration of TGFβ1-induced A549 cells in vitro. Gambogic acid also increased the mRNA and protein expression of E-cadherin, but repressed the mRNA and protein expression of N-cadherin, vimentin, and transcription factor TWIST1. Further examination of the mechanism revealed that the nuclear factor κB (NF-κB) pathway is involved in this regulation of EMT-related biomarkers. Gambogic acid inhibited NF-κB p65 nuclear translocation and the phosphorylation of the inhibitor of NF-κB (IκBα) and IκBα kinase (IKKα). Gambogic acid also suppressed the EMT induced by TGFβ1 and tumor necrosis factor α by inhibiting the NF-κB pathway. Our data also indicate that gambogic acid inhibited the primary lesion and lung metastasis of orthotopic model of A549 cells in vivo. We propose that gambogic acid might be developed as a candidate drug with therapeutic potential for the treatment of cancer invasion and migration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU078971 MISSION® esiRNA, esiRNA human TFRC (esiRNA1)
S0447
SIS3, ≥98% (HPLC), powder
C28H27N3O3 · HCl