EMAIL THIS PAGE TO A FRIEND
Scientific reports
Nanomechanics of the endothelial glycocalyx contribute to Na
PMID 28406245
Abstract
High dietary salt (NaCl) is a known risk factor for cardiovascular pathologies and inflammation. High plasma Na
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
208272
Vanadium(III) chloride, 97%
Cl3V