EMAIL THIS PAGE TO A FRIEND
Journal of controlled release : official journal of the Controlled Release Society
Interleukin-15 and cisplatin co-encapsulated thermosensitive polypeptide hydrogels for combined immuno-chemotherapy.
PMID 28408199
Abstract
In situ-forming thermosensitive hydrogels based on poly(ethylene glycol)-poly(γ-ethyl-l-glutamate) diblock copolymers (mPEG-b-PELG) were prepared for the co-delivery of interleukin-15 (IL-15) and cisplatin (CDDP). The polypeptide-based hydrogels as local drug delivery carriers could reduce the systemic toxicity, degrade thoroughly within 3weeks after subcutaneous injection into rats and display an acceptable biocompatibility. When incubated with mouse melanoma B16 cells, only the CDDP-treated groups had significant effects on the S phase cell-cycle arrest in melanoma cells. After a single peritumoral injection of the hydrogel containing IL-15/CDDP in C57BL/6 mice inoculated with B16F0-RFP melanoma cells, the dual drug-loaded hydrogels displayed synergistic anticancer efficacy, which was resulted from a combination of CDDP-mediated S arrest and IL-15/CDDP-induced recovery of CD8
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
673099
Biphenyl-4,4′-dithiol, 95%
C12H10S2