EMAIL THIS PAGE TO A FRIEND
The Journal of steroid biochemistry and molecular biology
Inhibition of heat shock protein 90 rescues glucocorticoid-induced bone loss through enhancing bone formation.
PMID 28408351
Abstract
Endogenous glucocorticoids (GCs) support normal bone development and bone mass maintenance, whereas long-term exposure to pharmacological dosages of GCs uncouples bone formation and resorption, resulting in GC-induced osteoporosis (GIOP). Heat shock protein 90 (HSP90) chaperoning glucocorticoid receptor (GR) signaling prompts us to speculate that HSP90 plays critical roles in GC-mediated bone formation and GIOP. In the present study, inhibition of HSP90 activity by 17-Demethoxy-17-allyaminogeldanmycin (17-AAG) or knockdown of HSP90 expression by siRNAs attenuated dexamethasone(Dex)-induced GR nuclear accumulation and transcriptional output of GR signaling, whereas overexpression of HSP90α or HSP90β enhanced GR transactivity in C3H10T1/2 cells. Though 17-AAG itself enhanced osteoblastic differentiation, it restored the Dex(10