EMAIL THIS PAGE TO A FRIEND
Diabetes
Renal and Vascular Effects of Uric Acid Lowering in Normouricemic Patients With Uncomplicated Type 1 Diabetes.
PMID 28408434
Abstract
Higher plasma uric acid (PUA) levels are associated with lower glomerular filtration rate (GFR) and higher blood pressure (BP) in patients with type 1 diabetes (T1D). Our aim was to determine the impact of PUA lowering on renal and vascular function in patients with uncomplicated T1D. T1D patients (n = 49) were studied under euglycemic and hyperglycemic conditions at baseline and after PUA lowering with febuxostat (FBX) for 8 weeks. Healthy control subjects were studied under normoglycemic conditions (n = 24). PUA, GFR (inulin), effective renal plasma flow (para-aminohippurate), BP, and hemodynamic responses to an infusion of angiotensin II (assessment of intrarenal renin-angiotensin-aldosterone system [RAAS]) were measured before and after FBX treatment. Arterial stiffness, flow-mediated dilation (FMD), nitroglycerin-mediated dilation (GMD), urinary nitric oxide (NO), and inflammatory markers were measured before and after FBX treatment. Gomez equations were used to estimate arteriolar afferent resistance, efferent resistance (RE), and glomerular hydrostatic pressure (PGLO). FBX had a modest systolic BP-lowering effect in T1D patients (112 ± 10 to 109 ± 9 mmHg, P = 0.049) without impacting arterial stiffness, FMD, GMD, or NO. FBX enhanced the filtration fraction response to hyperglycemia in T1D patients through larger increases in RE, PGLO, and interleukin-18 but without impacting the RAAS. FBX lowered systolic BP and modulated the renal RE responses to hyperglycemia but without impacting the RAAS or NO levels, suggesting that PUA may augment other hemodynamic or inflammatory mechanisms that control the renal response to hyperglycemia at the efferent arteriole. Ongoing outcome trials will determine cardiorenal outcomes of PUA lowering in patients with T1D.