EMAIL THIS PAGE TO A FRIEND
PMID 28408540
Abstract
Vitamin B
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M6546
(±)-2-(p-Methoxyphenoxy)propionic acid, ≥98%
C10H12O4
P5669
Pyridoxine, ≥98%
C8H11NO3