EMAIL THIS PAGE TO A FRIEND
Biotechnology and bioengineering
An in vitro synthetic biology platform for the industrial biomanufacturing of myo-inositol from starch.
PMID 28409846
Abstract
Myo-Inositol (vitamin B8) is widely used in the drug, cosmetic, and food & feed industries. Here, we present an in vitro non-fermentative enzymatic pathway that converts starch to inositol in one vessel. This in vitro pathway is comprised of four enzymes that operate without ATP or NAD
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
745480
Zinc isopropylsulfinate, 95%
C6H14O4S2Zn