Journal of pharmacological sciences
Permissive role of reduced inwardly-rectifying potassium current density in the automaticity of the guinea pig pulmonary vein myocardium.
PMID 28410965
The electrophysiological properties underlying the automaticity of the guinea pig pulmonary vein myocardium were studied. About 30% of the isolated pulmonary vein tissue preparations showed spontaneous electrical activity, as shown by glass microelectrode recordings from their myocardial layer. The remaining quiescent preparations had a resting membrane potential less negative than that in the left atria. Blockade of the acetylcholine activated potassium current (IK-ACh) by tertiapin induced a depolarizing shift of the resting membrane potential and automatic electrical activity in the pulmonary vein, but not in the atria. The tertiapin-induced electrical activity, as well as the spontaneous activity, was inhibited by the application of carbachol or by chelation of intracellular Ca(2+) by BAPTA. The isolated pulmonary vein cardiomyocytes had an IK-ACh density similar to that of the atrial cardiomyocytes, but a lower density of the inwardly-rectifying potassium current (IK1). Spontaneous Ca(2+) transients were observed in about 30% of the isolated pulmonary vein cardiomyocytes, but not in atrial cardiomyocytes. The Ca(2+) transients in the pulmonary vein cardiomyocytes were induced by tertiapin and inhibited by carbachol. These results indicate that the pulmonary vein cardiomyocytes have a reduced density of the inwardly-rectifying potassium current, which plays a permissive role in their intracellular Ca(2+)-dependent automaticity.
Related Materials
Product #
Molecular Formula
Add to Cart
E-4031, ≥98% (HPLC), solid
C21H27N3O3S · 2HCl