EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Closed-channel culture system for efficient and reproducible differentiation of human pluripotent stem cells into islet cells.
PMID 28412348
Abstract
Human pluripotent stem cells (hPSCs) are thought to be a promising cell-source solution for regenerative medicine due to their indefinite proliferative potential and ability to differentiate to functional somatic cells. However, issues remain with regard to achieving reproducible differentiation of cells with the required functionality for realizing human transplantation therapies and with regard to reducing the potential for bacterial or fungal contamination. To meet these needs, we have developed a closed-channel culture device and corresponding control system. Uniformly-sized spheroidal hPSCs aggregates were formed inside wells within a closed-channel and maintained continuously throughout the culture process. Functional islet-like endocrine cell aggregates were reproducibly induced following a 30-day differentiation protocol. Our system shows an easily scalable, novel method for inducing PSC differentiation with both purity and functionality.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S4572
SANT-1, ≥98% (HPLC), powder
C23H27N5