EMAIL THIS PAGE TO A FRIEND
ACS sustainable chemistry & engineering
Experimental Studies on the Hydrotreatment of Kraft Lignin to Aromatics and Alkylphenolics Using Economically Viable Fe-Based Catalysts.
PMID 28413733
Abstract
Limonite, a low-cost iron ore, was investigated as a potential hydrotreatment catalyst for kraft lignin without the use of an external solvent (batch reactor, initial H
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
71063
Goethite, 30-63% Fe
HFeO2