EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Investigation on the relationship between solubility of artemisinin and polyvinylpyrroli done addition by using DAOSD approach.
PMID 28414978
Abstract
In this work, we investigated the influence of polyvinylpyrrolidone (PVP) on the solubility of artemisinin in aqueous solution by using quantitative