EMAIL THIS PAGE TO A FRIEND
Oncotarget
Long noncoding RNA PCAT-14 induces proliferation and invasion by hepatocellular carcinoma cells by inducing methylation of miR-372.
PMID 28415780
Abstract
Long non-coding RNAs (lncRNAs) regulate oncogenesis by inducing methylation of CpG islands to silence target genes. Here we show that the lncRNA PCAT-14 is overexpressed in patients with hepatocellular carcinoma (HCC), and is associated with a poor prognosis after surgery. Our results demonstrate that PCAT-14 promotes proliferation, invasion, and cell cycle arrest in HCC cells. In addition, PCAT-14 inhibits miR-372 expression by inducing methylation of the miR-372 promoter. Simultaneously, miR-372 eliminates the effects of PCAT-14 on proliferation, invasion, and cell cycle in HCC cells. Moreover, PCAT-14 regulates expression of ATAD2 and activation of the Hedgehog pathway via miR-372. These findings indicate that PCAT-14 plays an important role in HCC, and may serve as a novel prognostic factor and therapeutic target.