EMAIL THIS PAGE TO A FRIEND
Oncotarget
Synergistic effects of the immune checkpoint inhibitor CTLA-4 combined with the growth inhibitor lycorine in a mouse model of renal cell carcinoma.
PMID 28416753
Abstract
Renal cell carcinoma (RCC) management has undergone a major transformation over the past decade; immune checkpoint inhibitors are currently undergoing clinical trials and show promising results. However, the effectiveness of immune checkpoint inhibitors in patients with metastatic RCC (mRCC) is still limited. Lycorine, an alkaloid extracted from plants of the Amaryllidaceae family, is touted as a potential anti-cancer drug because of its demonstrative growth inhibition capacity (induction of cell cycle arrest and inhibition of vasculogenic mimicry formation). Moreover, T cell checkpoint blockade therapy with antibodies targeting cytotoxic T-lymphocyte associated protein 4 (CTLA-4) has improved outcomes in cancer patients. However, the anti-tumor efficacy of combined lycorine and anti-CTLA-4 therapy remains unknown. Thus, we investigated a combination therapy of lycorine hydrochloride and anti-CTLA-4 using a murine RCC model. As a means of in vitro confirmation, we found that lycorine hydrochloride inhibited the viability of various RCC cell lines. Furthermore, luciferase-expressing Renca cells were implanted in the left kidney and the lung of BALB/c mice to develop a RCC metastatic mouse model. Lycorine hydrochloride and anti-CTLA-4 synergistically decreased tumor weight, lung metastasis, and luciferin-staining in tumor images. Importantly, the observed anti-tumor effects of this combination were dependent on significantly suppressing regulatory T cells while upregulating effector T cells; a decrease in regulatory T cells by 31.43% but an increase in effector T cells by 31.59% were observed in the combination group compared with those in the control group). We suggest that a combination of lycorine hydrochloride and anti-CTLA-4 is a viable therapeutic option for RCC patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L5139
Lycorine hydrochloride, ≥98% (TLC), powder
C16H17NO4 · HCl